Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XTH31
Entrez Gene ID
823634
Gene Name
xyloglucan endo-transglycosylase-related 8
Gene Aliases
AT3G44990, ATXTR8, AtXTH31, XTR8, XYLOGLUCAN ENDOTRANSGLUCOSYLASE/HYDROLASE 31, xyloglucan endo-transglycosylase-related 8
Chromosome Location
Chr.3: 16446935 - 16448844 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114368.3 NP_190085.1 3 - 4 684 150
GenBank mRNA AY056163.1 - 3 - 4 604 150
AY136454.1 - 3 - 4 604 150
BT006326.1 - 3 - 4 518 150

View Full Product Details