Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRK2A
Entrez Gene ID
815274
Gene Name
Leucine-rich repeat protein kinase family protein
Gene Aliases
AT2G07040, ATPRK2A, AtPRK2, PRK2, T4E14.15, T4E14_15, pollen receptor like kinase 2
Chromosome Location
Chr.2: 2916416 - 2919392 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126680.4 NP_178721.3 2 - 3 1481 99
GenBank mRNA BT003936.1 - 2 - 3 1359 99
BT005118.1 - 2 - 3 1276 99
FJ708690.1 - 2 - 3 1276 99

View Full Product Details