Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAK10
Entrez Gene ID
815566
Gene Name
MAK10 homologue
Gene Aliases
AT2G11000, ATMAK10, F15K19.7, F15K19_7, MAK10 homologue
Chromosome Location
Chr.2: 4341105 - 4347974 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001124823.2 NP_001118295.1 3 - 4 553 109
NM_001335364.1 NP_001324918.1 3 - 4 483 109
NM_001335365.1 NP_001324917.1 3 - 4 447 109
NM_126831.4 NP_178872.2 3 - 4 565 109

View Full Product Details