Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHR5
Entrez Gene ID
815823
Gene Name
chromatin remodeling 5
Gene Aliases
AT2G13370, F14O4.6, F14O4_6, chromatin remodeling 5
Chromosome Location
Chr.2: 5544054 - 5556560 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335389.1 NP_001325087.1 28 - 29 5201 155
NM_001335390.1 NP_001325086.1 28 - 29 4597 155
NM_126926.5 NP_178970.3 28 - 29 4795 155

View Full Product Details