Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NTH2
Entrez Gene ID
837100
Gene Name
endonuclease III 2
Gene Aliases
AT1G05900, ATNTH2, T20M3.18, T20M3_18, endonuclease III 2
Chromosome Location
Chr.1: 1786849 - 1789737 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100471.4 NP_563752.1 9 - 10 1188 65
NM_202038.4 NP_973767.1 10 - 11 1059 65
GenBank mRNA AF410268.1 - 9 - 10 1171 65
AK316958.1 - 10 - 11 1052 65

View Full Product Details