Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBOHB
Entrez Gene ID
837430
Gene Name
respiratory burst oxidase-like protein
Gene Aliases
AT1G09090, ATRBOHB, ATRBOHB-BETA, F7G19.4, F7G19_4, respiratory burst oxidase homolog B
Chromosome Location
Chr.1: 2932739 - 2936586 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100780.3 NP_172383.3 9 - 10 2007 81
NM_202070.1 NP_973799.1 10 - 11 1933 81
GenBank mRNA BT005716.1 - 9 - 10 2008 81

View Full Product Details