Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBOHB
Entrez Gene ID
837430
Gene Name
respiratory burst oxidase-like protein
Gene Aliases
AT1G09090, ATRBOHB, ATRBOHB-BETA, F7G19.4, F7G19_4, respiratory burst oxidase homolog B
Chromosome Location
Chr.1: 2932739 - 2936586 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100780.3 NP_172383.3 4 - 5 706 77
NM_202070.1 NP_973799.1 4 - 5 706 77
GenBank mRNA BT005716.1 - 4 - 5 707 77

View Full Product Details