Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDX15
Entrez Gene ID
839773
Gene Name
nudix hydrolase homolog 15
Gene Aliases
ARABIDOPSIS THALIANA NUDIX HYDROLASE HOMOLOG 15, AT1G28960, ATNUDT15, ATNUDX15, F1K23.5, F1K23_5, nudix hydrolase homolog 15
Chromosome Location
Chr.1: 10109500 - 10111738 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036027.2 NP_001031104.1 2 - 3 661 91
NM_001332826.1 NP_001320887.1 2 - 3 654 91
NM_102640.3 NP_564316.1 2 - 3 661 91
NM_179393.4 NP_849724.1 2 - 3 661 91
NM_179394.3 NP_849725.2 2 - 3 661 91
GenBank mRNA AK175332.1 - 2 - 3 571 91
AK176465.1 - 2 - 3 573 91
AK221329.1 - 2 - 3 536 91
AK228486.1 - 2 - 3 583 91
AK230433.1 - 2 - 3 536 91
AK317052.1 - 2 - 3 565 91

View Full Product Details