Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLPS3
Entrez Gene ID
819626
Gene Name
CLP-similar protein 3
Gene Aliases
AT3G04680, CLP-similar protein 3, F7O18.15, F7O18_15
Chromosome Location
Chr.3: 1269722 - 1273004 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337539.1 NP_001319471.1 2 - 3 491 73
NM_001337540.1 NP_001327497.1 1 - 2 739 73
NM_111340.3 NP_187119.1 2 - 3 528 73
GenBank mRNA AY062866.1 - 2 - 3 486 73
AY128751.1 - 1 - 2 220 73

View Full Product Details