Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NHX2
Entrez Gene ID
819665
Gene Name
sodium hydrogen exchanger 2
Gene Aliases
AT3G05030, ATNHX2, T9J14.2, T9J14_2, sodium hydrogen exchanger 2
Chromosome Location
Chr.3: 1393142 - 1397627 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337553.1 NP_001319475.1 10 - 11 1278 129
NM_001337554.1 NP_001326211.1 10 - 11 1525 129
NM_001337555.1 NP_001326210.1 9 - 10 1696 129
NM_111375.4 NP_187154.1 10 - 11 1525 129
GenBank mRNA AF490586.1 - 9 - 10 909 129

View Full Product Details