Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACBP4
Entrez Gene ID
819707
Gene Name
acyl-CoA binding protein 4
Gene Aliases
AT3G05420, AtACBP4, F22F7.13, F22F7_13, acyl-CoA binding protein 4
Chromosome Location
Chr.3: 1561671 - 1567336 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111415.5 NP_187193.3 8 - 9 920 95
NM_202498.2 NP_974227.1 8 - 9 1030 95
GenBank mRNA AK228046.1 - 8 - 9 921 95
BT006458.1 - 8 - 9 818 95

View Full Product Details