Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LACS6
Entrez Gene ID
819767
Gene Name
long-chain acyl-CoA synthetase 6
Gene Aliases
AT3G05970, ATLACS6, F2O10.7, F2O10_7, long-chain acyl-CoA synthetase 6
Chromosome Location
Chr.3: 1786279 - 1791887 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111471.3 NP_566265.1 16 - 17 1569 84
GenBank mRNA AY094418.1 - 16 - 17 1483 84
BT001117.1 - 16 - 17 1428 84

View Full Product Details