Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NP3
Entrez Gene ID
819774
Gene Name
NPK1-related protein kinase 3
Gene Aliases
ANP3, AT3G06030, AtANP3, F24F17.1, F24F17_1, MAPKKK12, NPK1-related protein kinase 3
Chromosome Location
Chr.3: 1818732 - 1822858 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111477.4 NP_187254.1 6 - 7 821 70
GenBank mRNA AB000799.1 - 6 - 7 709 70
AY069917.1 - 6 - 7 803 70
BT001024.1 - 6 - 7 668 70

View Full Product Details