Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MEE32
- Entrez Gene ID
-
819809
- Gene Name
- dehydroquinate dehydratase, putative / shikimate dehydrogenase
- Gene Aliases
-
AT3G06350, EMB3004, EMBRYO DEFECTIVE 3004, F24P17.18, F24P17_18, MATERNAL EFFECT EMBRYO ARREST 32
- Chromosome Location
-
Chr.3: 1924270 - 1927850 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_111510.4 |
NP_187286.1 |
8 - 9 |
1610 |
140 |
View Full Product Details