Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NHX4
Entrez Gene ID
819811
Gene Name
sodium hydrogen exchanger 4
Gene Aliases
AT3G06370, ATNHX4, F24P17.16, F24P17_16, sodium hydrogen exchanger 4
Chromosome Location
Chr.3: 1929946 - 1934073 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337650.1 NP_001327378.1 2 - 3 371 72
NM_001337651.1 NP_001327379.1 2 - 3 371 72
NM_001337652.1 NP_001327377.1 1 - 2 301 72
NM_111512.3 NP_187288.2 2 - 3 371 72

View Full Product Details