Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSL10
Entrez Gene ID
819903
Gene Name
glucan synthase-like 10
Gene Aliases
AT3G07160, ATGSL10, CALS9, glucan synthase-like 10, gsl10
Chromosome Location
Chr.3: 2264862 - 2279916 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337693.1 NP_001326712.1 40 - 41 4426 84
NM_001337694.1 NP_001326713.1 39 - 40 4294 84
NM_111596.6 NP_187372.5 39 - 40 4230 84

View Full Product Details