Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHIP
Entrez Gene ID
819925
Gene Name
carboxyl terminus of HSC70-interacting protein
Gene Aliases
AT3G07370, ATCHIP, CARBOXYL TERMINUS OF HSC70-INTERACTING PROTEIN, carboxyl terminus of HSC70-interacting protein
Chromosome Location
Chr.3: 2357907 - 2360549 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111616.3 NP_566305.1 2 - 3 417 65
GenBank mRNA AY042807.1 - 2 - 3 417 65
AY064647.1 - 2 - 3 169 65

View Full Product Details