Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYB83
Entrez Gene ID
819997
Gene Name
myb domain protein 83
Gene Aliases
AT3G08500, AtMYB83, myb domain protein 83
Chromosome Location
Chr.3: 2576444 - 2578276 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111685.3 NP_187463.1 1 - 2 528 67
GenBank mRNA AB493606.1 - 1 - 2 324 67
AF371974.1 - 1 - 2 324 67
BT026506.1 - 1 - 2 324 67
DQ446645.1 - 1 - 2 324 67

View Full Product Details