Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLC2
Entrez Gene ID
819999
Gene Name
phospholipase C 2
Gene Aliases
AT3G08510, ATPLC2, phospholipase C 2
Chromosome Location
Chr.3: 2582362 - 2585898 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035583.1 NP_001030660.1 2 - 3 788 75
NM_001161124.1 NP_001154596.1 2 - 3 788 75
NM_001337760.1 NP_001327169.1 2 - 3 788 75
NM_111686.6 NP_187464.1 2 - 3 835 75
GenBank mRNA AF360206.1 - 2 - 3 556 75
AK318857.1 - 2 - 3 646 75
AY040054.1 - 2 - 3 493 75
D50804.1 - 2 - 3 667 75

View Full Product Details