Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- YSL1
- Entrez Gene ID
-
828512
- Gene Name
- YELLOW STRIPE like 1
- Gene Aliases
-
AT4G24120, ATYSL1, T19F6.110, T19F6_110, YELLOW STRIPE LIKE 1, YELLOW STRIPE like 1
- Chromosome Location
-
Chr.4: 12524186 - 12527347 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118544.4 |
NP_567694.2 |
2 - 3 |
1054 |
89 |
View Full Product Details