Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHD
Entrez Gene ID
828520
Gene Name
Chaperone protein htpG family protein
Gene Aliases
AT4G24190, AtHsp90-7, AtHsp90.7, HEAT SHOCK PROTEIN 90-7, HEAT SHOCK PROTEIN 90.7, HSP90.7, SHEPHERD, T22A6.20, T22A6_20
Chromosome Location
Chr.4: 12551552 - 12556058 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118552.4 NP_194150.1 8 - 9 1114 155
NM_202877.1 NP_974606.1 8 - 9 961 155
GenBank mRNA AB064528.1 - 8 - 9 947 155
AY039895.1 - 8 - 9 955 155
BT004527.1 - 8 - 9 907 155

View Full Product Details