Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MC2
Entrez Gene ID
828614
Gene Name
metacaspase 2
Gene Aliases
AT4G25110, AtMC2, AtMCP1c, F24A6.7, MCP1c, metacaspase 1c, metacaspase 2
Chromosome Location
Chr.4: 12887474 - 12890262 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036634.1 NP_001031711.1 2 - 3 1122 89
NM_118643.5 NP_194241.3 2 - 3 1126 89
GenBank mRNA AK175293.1 - 2 - 3 933 89
AK221790.1 - 2 - 3 1126 89
AY219827.1 - 2 - 3 817 89
BT026456.1 - 2 - 3 817 89

View Full Product Details