Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCB28
Entrez Gene ID
828650
Gene Name
non-intrinsic ABC protein 8
Gene Aliases
ARABIDOPSIS THALIANA NON-INTRINSIC ABC PROTEIN 8, AT4G25450, AT4G25460, ATNAP8, ATP-binding cassette B28, NAP8, T30C3.5, non-intrinsic ABC protein 8
Chromosome Location
Chr.4: 13009690 - 13014085 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036637.2 NP_001031714.1 1 - 2 446 83
NM_118677.5 NP_194275.2 1 - 2 560 83
GenBank mRNA AK175486.1 - 1 - 2 444 83
AY094470.1 - 1 - 2 441 83

View Full Product Details