Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSN6B
Entrez Gene ID
828749
Gene Name
COP9 signalosome subunit 6B
Gene Aliases
AT4G26430, COP9 signalosome subunit 6B, M3E9.140, M3E9_140
Chromosome Location
Chr.4: 13354992 - 13357629 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118776.3 NP_567746.1 3 - 4 716 82
GenBank mRNA AF395064.1 - 3 - 4 479 82
AY087556.1 - 3 - 4 486 82
BT002345.1 - 3 - 4 479 82

View Full Product Details