Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ATS1
- Entrez Gene ID
-
828781
- Gene Name
- peroxygenase 1
- Gene Aliases
-
ARABIDOPSIS THALIANA PEROXYGENASE 1, AT4G26740, ATPXG1, AtCLO1, CALEOSIN1, CLO1, F10M23.80, F10M23_80, seed gene 1
- Chromosome Location
-
Chr.4: 13473516 - 13475371 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118808.5 |
NP_194404.1 |
1 - 2 |
211 |
71 |
View Full Product Details