Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP71A23
Entrez Gene ID
823988
Gene Name
cytochrome P450, family 71, subfamily A, polypeptide 23
Gene Aliases
''cytochrome P450, AT3G48300, cytochrome P450, family 71, polypeptide 23, polypeptide 23'', subfamily A
Chromosome Location
Chr.3: 17885465 - 17887272 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339360.1 NP_001325471.1 1 - 2 911 78
NM_148856.3 NP_680109.2 1 - 2 852 78

View Full Product Details