Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REF6
Entrez Gene ID
824002
Gene Name
relative of early flowering 6
Gene Aliases
AT3G48430, JMJ12, Jumonji domain-containing protein 12, relative of early flowering 6
Chromosome Location
Chr.3: 17935256 - 17940792 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148863.4 NP_680116.2 3 - 4 1377 69
GenBank mRNA AY664499.1 - 3 - 4 1024 69

View Full Product Details