Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATKRS-1
Entrez Gene ID
820343
Gene Name
lysyl-tRNA synthetase 1
Gene Aliases
AT3G11710, lysyl-tRNA synthetase 1
Chromosome Location
Chr.3: 3702048 - 3705716 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112003.4 NP_187777.1 5 - 6 621 91
GenBank mRNA AF125574.1 - 5 - 6 604 91
AY099551.1 - 5 - 6 582 91
BT002123.1 - 5 - 6 518 91

View Full Product Details