Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HCHIB
- Entrez Gene ID
-
820429
- Gene Name
- basic chitinase
- Gene Aliases
-
AT3G12500, ATHCHIB, B-CHI, CHI-B, PATHOGENESIS-RELATED 3, PR-3, PR3, basic chitinase
- Chromosome Location
-
Chr.3: 3962136 - 3963984 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112085.4 |
NP_566426.2 |
1 - 2 |
462 |
62 |
View Full Product Details