Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ACA10
- Entrez Gene ID
-
829112
- Gene Name
- autoinhibited Ca(2+)-ATPase 10
- Gene Aliases
-
AT4G29900, ATACA10, AUTOINHIBITED CA(2+)-ATPASE, CIF1, COMPACT INFLORESCENCE 1, F27B13.140, F27B13_140, autoinhibited Ca(2+)-ATPase 10
- Chromosome Location
-
Chr.4: 14610883 - 14619685 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119136.4 |
NP_194719.1 |
34 - 35 |
3527 |
125 |
View Full Product Details