Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VLN4
Entrez Gene ID
829139
Gene Name
villin 4
Gene Aliases
AT4G30160, ATVLN4, F9N11.10, F9N11_10, villin 4
Chromosome Location
Chr.4: 14753232 - 14760236 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203940.1 NP_001190869.1 14 - 15 1879 105
NM_001341999.1 NP_001329918.1 14 - 15 1947 105
NM_001342000.1 NP_001329917.1 13 - 14 1877 105
NM_119162.5 NP_194745.1 13 - 14 2219 105
GenBank mRNA AK117296.1 - 13 - 14 1826 105
BT005980.1 - 12 - 13 1616 105
Y12782.1 - - 1859 105

View Full Product Details