Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VLN4
Entrez Gene ID
829139
Gene Name
villin 4
Gene Aliases
AT4G30160, ATVLN4, F9N11.10, F9N11_10, villin 4
Chromosome Location
Chr.4: 14753232 - 14760236 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203940.1 NP_001190869.1 15 - 16 2012 65
NM_001341999.1 NP_001329918.1 15 - 16 2080 65
NM_001342000.1 NP_001329917.1 14 - 15 2010 65
NM_119162.5 NP_194745.1 14 - 15 2352 65
GenBank mRNA AK117296.1 - 14 - 15 1959 65
BT005980.1 - 13 - 14 1749 65
Y12782.1 - - 1992 65

View Full Product Details