Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VLN4
Entrez Gene ID
829139
Gene Name
villin 4
Gene Aliases
AT4G30160, ATVLN4, F9N11.10, F9N11_10, villin 4
Chromosome Location
Chr.4: 14753232 - 14760236 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341999.1 NP_001329918.1 23 - 24 3287 120
NM_001342000.1 NP_001329917.1 22 - 23 3217 120
NM_119162.5 NP_194745.1 22 - 23 3559 120
GenBank mRNA AK117296.1 - 22 - 23 3166 120
Y12782.1 - - 3199 120

View Full Product Details