Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IAMT1
Entrez Gene ID
835618
Gene Name
IAA carboxylmethyltransferase 1
Gene Aliases
AT5G55250, AtIAMT1, IAA carboxylmethyltransferase 1, MCO15.20, MCO15_20
Chromosome Location
Chr.5: 22407427 - 22410974 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124907.5 NP_200336.1 1 - 2 232 102
GenBank mRNA AK175586.1 - 1 - 2 176 102

View Full Product Details