Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NHX3
- Entrez Gene ID
-
835640
- Gene Name
- Na+/H+ (sodium hydrogen) exchanger 3
- Gene Aliases
-
ARABIDOPSIS THALIANA NA+/H+ (SODIUM HYDROGEN) EXCHANGER 3, AT5G55470, ATNHX3, MTE17.18, MTE17_18, Na+/H+ (sodium hydrogen) exchanger 3
- Chromosome Location
-
Chr.5: 22469245 - 22472706 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124929.3 |
NP_200358.1 |
6 - 7 |
803 |
98 |
GenBank mRNA |
AK221571.1 |
- |
6 - 7 |
803 |
98 |
View Full Product Details