Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SVL1
Entrez Gene ID
835641
Gene Name
SHV3-like 1
Gene Aliases
AT5G55480, GDPDL4, GPDL1, Glycerophosphodiester phosphodiesterase (GDPD) like 4, MTE17.19, MTE17_19, SHV3-like 1, glycerophosphodiesterase-like 1
Chromosome Location
Chr.5: 22474142 - 22478216 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124930.4 NP_200359.1 5 - 6 1223 85
GenBank mRNA BT015819.1 - 5 - 6 1135 85
BT020207.1 - 5 - 6 1088 85

View Full Product Details