Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT4G19110
Entrez Gene ID
827649
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT4G19110, T18B16.80, T18B16_80
Chromosome Location
Chr.4: 10454497 - 10459328 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084939.1 NP_001078408.1 7 - 8 579 78
NM_118030.3 NP_567574.1 11 - 12 1651 78
NM_179076.1 NP_849407.1 11 - 12 1621 78
GenBank mRNA AF360190.1 - 11 - 12 1595 78
AY040004.1 - 8 - 9 713 78

View Full Product Details