Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT4G19110
Entrez Gene ID
827649
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT4G19110, T18B16.80, T18B16_80
Chromosome Location
Chr.4: 10454497 - 10459328 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179076.1 NP_849407.1 15 - 16 2047 67

View Full Product Details