Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT10
Entrez Gene ID
828648
Gene Name
nudix hydrolase homolog 10
Gene Aliases
AT4G25434, ATNUDT10, nudix hydrolase homolog 10
Chromosome Location
Chr.4: 13003568 - 13005935 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036636.2 NP_001031713.1 6 - 7 882 99
NM_001341732.1 NP_001329739.1 7 - 8 780 99
NM_001341733.1 NP_001329738.1 5 - 6 925 99
NM_001341734.1 NP_001329741.1 6 - 7 844 99
NM_001341735.1 NP_001329740.1 5 - 6 991 99
NM_001341736.1 NP_001329746.1 5 - 6 1170 99
NM_001341737.1 NP_001329743.1 5 - 6 1155 99
NM_001341738.1 NP_001329742.1 6 - 7 1076 99
NM_001341741.1 NP_001329748.1 6 - 7 861 99
NM_179112.3 NP_849443.2 7 - 8 765 99
GenBank mRNA BT010765.1 - 7 - 8 729 99
BT010987.1 - 7 - 8 637 99

View Full Product Details