Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAMMA-ADAPTIN 1
Entrez Gene ID
839001
Gene Name
gamma-adaptin 1
Gene Aliases
AT1G23900, Gamma-ADR, Gamma-adaptin, gamma-adaptin 1
Chromosome Location
Chr.1: 8441073 - 8447681 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332624.1 NP_001322140.1 7 - 8 1635 83
NM_102238.2 NP_173802.1 8 - 9 1215 83
NM_179370.4 NP_849701.1 7 - 8 1372 83
GenBank mRNA BT004307.1 - 7 - 8 1288 83
BT005557.1 - 7 - 8 1072 83

View Full Product Details