Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATJ1
Entrez Gene ID
839715
Gene Name
DNAJ heat shock family protein
Gene Aliases
AT1G28210, ATJ, AtDjB1, F3H9.13, F3H9_13
Chromosome Location
Chr.1: 9854517 - 9860158 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332806.1 NP_001321133.1 11 - 12 777 82
NM_102586.3 NP_174142.1 11 - 12 777 82
NM_179388.2 NP_849719.1 11 - 12 777 82

View Full Product Details