Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCC6
Entrez Gene ID
820497
Gene Name
multidrug resistance-associated protein 8
Gene Aliases
AT3G13090, ATABCC6, ATMRP8, ATP-binding cassette C6, MRP8, multidrug resistance-associated protein 8
Chromosome Location
Chr.3: 4202811 - 4208261 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112148.4 NP_187916.3 8 - 9 4379 63

View Full Product Details