Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLIP4
Entrez Gene ID
820641
Gene Name
GDSL-motif lipase 4
Gene Aliases
AT3G14225, GDSL-motif lipase 4
Chromosome Location
Chr.3: 4734553 - 4735993 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338115.1 NP_001325624.1 1 - 2 269 68
NM_112280.1 NP_188039.1 1 - 2 269 68
GenBank mRNA MF774103.1 - 1 - 2 269 68

View Full Product Details