Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSL04
Entrez Gene ID
820683
Gene Name
glucan synthase-like 4
Gene Aliases
AT3G14570, ATGSL04, GSL4, atgsl4, glucan synthase-like 4, gsl04
Chromosome Location
Chr.3: 4892410 - 4902880 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202964.2 NP_001189893.1 30 - 31 3525 85
NM_001338124.1 NP_001326811.1 29 - 30 3675 85
NM_112317.3 NP_188075.2 29 - 30 3762 85

View Full Product Details