Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WIN2
Entrez Gene ID
829303
Gene Name
HOPW1-1-interacting 2
Gene Aliases
AT4G31750, F28M20.60, F28M20_60, HOPW1-1-interacting 2
Chromosome Location
Chr.4: 15363708 - 15367892 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342110.1 NP_001328539.1 5 - 6 899 91
NM_001342111.1 NP_001328538.1 5 - 6 943 91
NM_119324.4 NP_194903.2 5 - 6 768 91
GenBank mRNA AY080658.1 - 5 - 6 721 91
AY133761.1 - 4 - 5 460 91
EU214909.1 - 4 - 5 460 91

View Full Product Details