Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PANK2
Entrez Gene ID
829351
Gene Name
pantothenate kinase 2
Gene Aliases
AT4G32180, ATPANK2, F10M6.180, F10M6_180, pantothenate kinase 2
Chromosome Location
Chr.4: 15537559 - 15543954 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036691.1 NP_001031768.1 16 - 17 2145 124
NM_001125620.1 NP_001119092.1 16 - 17 2145 124
NM_119370.5 NP_194945.3 16 - 17 2195 124

View Full Product Details