Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIFY8
Entrez Gene ID
829392
Gene Name
TIFY domain protein 8
Gene Aliases
AT4G32570, F4D11.230, F4D11_230, TIFY domain protein 8
Chromosome Location
Chr.4: 15716232 - 15718867 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342158.1 NP_001328277.1 1 - 2 401 85
NM_119409.4 NP_567898.1 1 - 2 410 85
GenBank mRNA BT006431.1 - 1 - 2 245 85

View Full Product Details