Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGT73B2
Entrez Gene ID
829560
Gene Name
UDP-glucosyltransferase 73B2
Gene Aliases
AT4G34135, UDP-glucosyltransferase 73B2
Chromosome Location
Chr.4: 16345150 - 16347434 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119575.2 NP_567954.1 1 - 2 1331 76
GenBank mRNA AY035164.1 - 1 - 2 1100 76
AY142692.1 - 1 - 2 983 76
AY339370.1 - 1 - 2 1098 76

View Full Product Details