Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GBF6
Entrez Gene ID
829611
Gene Name
G-box binding factor 6
Gene Aliases
AT4G34590, ATB2, ATBZIP11, Arabidopsis thaliana basic leucine-zipper 11, AtbZIP11, BZIP11, G-box binding factor 6, T4L20.170, T4L20_170
Chromosome Location
Chr.4: 16521861 - 16523230 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119625.3 NP_195185.1 1 - 1 1142 144
GenBank mRNA AY063979.1 - 1 - 1 1094 144

View Full Product Details