Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GGH1
Entrez Gene ID
844202
Gene Name
gamma-glutamyl hydrolase 1
Gene Aliases
AT1G78660, ATGGH1, T30F21.1, T30F21_1, gamma-glutamyl hydrolase 1
Chromosome Location
Chr.1: 29585677 - 29588389 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106513.5 NP_177987.1 6 - 7 769 147
NM_179569.4 NP_849900.1 5 - 6 740 147
NM_202443.3 NP_974172.1 7 - 8 800 147
GenBank mRNA AK317198.1 - 5 - 6 726 147
BT024737.1 - 5 - 6 662 147

View Full Product Details